missing translation for 'onlineSavingsMsg'
Learn More

Logistik Unicorp™ Faithful Spruce Green Color 65% Polyester/35% Cotton Lab Coats

Brand:  Logistik Unicorp™ 441-V6SG-96

Code : 92

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12871121

  • £40.40 / Each


Estimated Shipment
04-06-2018
Add to basket

Specifications

Specifications

Used for general laboratory safety
245g/m2
Lab Coat
One breast pocket, two lower patch pockets, centre back vent
96cm
Videos
Documentation

Technical Documents