missing translation for 'onlineSavingsMsg'
Learn More

Logistik Unicorp™ Faithful Navy Color Polyester/Cotton Lab Coats

Brand:  Logistik Unicorp™ 441YH-V6NA-116

UNSPSC : 46181532

Code : 92

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12851368

  • £43.35 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications Description & Specifications

Specifications

116cm
245g/m2
Two lower pockets and wrap over style stud fastened front
Lab Coat
Used for general laboratory safety