missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Factor XIIIa Mouse, Clone: SPM180, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 2162-MSM2X-P1
Additional Details : Weight : 0.01000kg
Specifications
Factor XIIIa | |
Cytoplasmic and Secreted | |
SPM180 | |
Unconjugated | |
NP_000120 | |
Mouse | |
83kDa | |
Protein A/G Purified | |
RUO | |
2162 | |
IgG2b κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
F13A1 | |
Recombinant fragment of human Factor XIIIa protein (aa46-181) | |
293T Cells. Placenta. | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |