missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitritex™ Fabric Coveralls With Hood
Click to view available options
Size:
Small
Medium
Large
Description
- Low-linting & durable CleanTough™material
- Thumb loops to ensure a secure hold
- Zip with sealable cover
- Elasticated hood, back, cuffs and ankles
- Tested against permeation standard ASTM D6978-05
Specifications
Product Type | Cleanroom Coveralls |
Size | Small |
Material | Polypropylene |
Color | White |
Disposable | Single-use |
Ankle Style | Elastic |
Cuff Style | Elastic |
Neck Style | Hooded |
Hood Type | Elastic |
Closure Type | Zipper |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'