Learn More
Thermo Scientific™ FABP4 Recombinant Mouse Protein, His Tag, mammalian expression system
Expressed from DNA sequence encoding mouse FABP4 (Met 1-Ala 132) fused to polyhistidine tag at C-terminus
Brand: Thermo Scientific™ 50652M08H5
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Formulation: 50mM Tris, 20% glycerol, pH 9.5
Specifications
Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications | |
11770 | |
>85% by SDS-PAGE | |
Mouse | |
<1 EU/μg | |
FABP4 | |
Other Proteins | |
FABP4 Recombinant Mouse Protein |
Lyophilized | |
16kd | |
5μg | |
Full Length, Recombinant | |
Fatty Acid Binding Protein 4 adipocyte | |
Assay Standard | |
Mammalian | |
His Tag |
For Research Use Only. Not for use in diagnostic procedures.