missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FABP4/A-FABP Antibody (FABP4/8537R) [Janelia Fluor™ 669], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-20999JF669
This item is not returnable.
View return policy
Description
FABP4/A-FABP Monoclonal antibody specifically detects FABP4/A-FABP in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| FABP4/A-FABP | |
| Monoclonal | |
| Janelia Fluor 669 | |
| Adipocyte lipid-binding protein, Adipocyte-type fatty acid-binding protein, A-FABPAFABP, ALBP, aP2, fatty acid binding protein 4, adipocyte, Fatty acid-binding protein 4, fatty acid-binding protein, adipocyte | |
| Recombinant fragment (around aa1-132) of human FABP4/A-FABP protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cancer, Phospho Specific, Stem Cell Markers | |
| 2167 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| FABP4/8537R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction