missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ FABP3 Recombinant Human Protein

Expressed from DNA sequence encoding human FABP3 (Met 1-Ala 133)

Brand:  Sino Biological™ 12476HNAE250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15255945

  • £363.31 / 250µg


EU Stock 2
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

  • Formulation: 50mM Tris, pH 8.2, 5% Mannitol, 5% Trehalose, 0.02% Tween80, 5% Mannitol, 5% Trehalose, 0.02% Tween80
Specifications

Specifications

Analysis of protein stucture and used in cell biology research applications
2170
≥90% by SDS-PAGE
Human
Fatty Acid Binding Protein 3, muscle and heart (mammary-derived growth inhibitor)
Assay Standard
E.coli
Untagged
Lyophilized
14.9kd
5 x 50μg
Full Length, Recombinant
FABP3
Other Proteins
FABP3 Recombinant Human Protein
Videos