missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ FABP3 Recombinant Human Protein

Expressed from DNA sequence encoding human FABP3 (Met 1-Ala 133)

Brand:  Sino Biological™ 12476HNAE50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15275945

  • £214.45 / 50µg


Estimated Shipment
15-06-2018
Add to basket

Description

Description

  • Formulation: 50mM Tris, pH 8.2, 5% Mannitol, 5% Trehalose, 0.02% Tween80, 5% Mannitol, 5% Trehalose, 0.02% Tween80
Specifications

Specifications

Analysis of protein stucture and used in cell biology research applications
2170
≥90% by SDS-PAGE
Human
Fatty Acid Binding Protein 3, muscle and heart (mammary-derived growth inhibitor)
Assay Standard
E.coli
Untagged
Lyophilized
14.9kd
50μg
Full Length, Recombinant
FABP3
Other Proteins
FABP3 Recombinant Human Protein
Videos

For Research Use Only. Not for use in diagnostic procedures.