missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ FABP2 Recombinant Human Protein, His Tag

Expressed from DNA sequence encoding human FABP2 (Met 1-Asp 132) fused to a polyhistidine tag at C-terminus

Brand:  Sino Biological™ 10130H08E50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15284705

  • £161.36 / 50µg


Estimated Shipment
02-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

  • Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
Specifications

Specifications

Lyophilized
16.6kd
50μg
−20°C
Fatty Acid Binding Protein 2, intestinal
Assay Standard
E.coli
His Tag
2169
≥95% by SDS-PAGE
Human
<1 EU/μg
FABP2
Other proteins
FABP2 Recombinant Human Protein
Videos