missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ FABP1 (L-FABP) Recombinant Human Protein, N-His Tag

Expressed from DNA sequence encoding human FABP1 (Ser 2-Ile 127) fused to polyhistide tag at N-terminus

Brand:  Sino Biological™ 12353H07E25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15235895

  • £81.98 / 25µg


EU Stock 5
Add to basket

Description

Description

  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH: 8.3, 5% Mannitol, 5% Trehalose, 0.02% Tween80
Specifications

Specifications

Analysis of protein stucture and used in cell biology research applications
2168
≥90% by SDS-PAGE
Human
Fatty Acid Binding Protein 1, liver
Assay Standard
E.coli
His Tag
Lyophilized
15.6kd
5 x 5μg
Full Length, Recombinant
FABP1
Other Proteins
FABP1 (L-FABP) Recombinant Human Protein
Videos

For Research Use Only. Not for use in diagnostic procedures.