Learn More
F4/80 Antigen Rat, V450, Clone: BM8.1, enQuireBio™
Rat Monoclonal Antibody
Brand:  EnquireBio QAB68-V450-100ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15952368
  • £360.04 / 100µg
Estimated Shipment: 17-01-2022
Specifications
F4/80 Antigen
BM8.1
V450
XP_006523664
Rat
Protein A/G Purified
RUO
Primary
13733
Flow Cytometry
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2
Adgre1
IgG2b kappa
100 μg
Store at 2-4°C.