missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ F1-ClipTip™ Good Laboratory Pipetting (GLP) Kits

For your convenience, we packaged a variety of F1-ClipTip™ pipettes in a range of volumes, compatible ClipTip pipette tips and a pipette stand to support good laboratory pipetting in your lab.

Brand:  Thermo Scientific™ 4701150N

Additional Details : Weight : 3.36000kg

 View more versions of this product

Product Code. 15591945

  • £942.24 / Each


Estimated Shipment
30-10-2020
Add to basket

Description

Description

Worry-free pipetting is finally here with the only tip that clips.
Sealed in security of ClipTip Technology. Feel the difference.


Eliminate the frustration of banging tips on your pipette
With a light touch, each tip is clipped securely on the pipette.

Reduce the risk of loose or leaking tips

When ClipTip pipette tips are on, they are sealed and will not loosen. The ClipTip pipetting system remains sealed until you decide to eject tips, even in the most comprehensive applications, including multiple tip touches and mixing cycles.

Enable consistent and reproducible pipetting from user to user

Higher-quality results and more efficient research.

Learn more >

Ergonomically In-touch
Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of RSI

Silent, Effortless Volume Adjustment
Enjoy the new silent and lighter volume adjustment mechanism with over 50% lower forces compared to earlier models.

Secure Volume Lock
The secure volume setting prevents undesirable volume drift during operation. Simply lift the plunger up to set volume and press down to lock in place with minimum effort.

Adjustable Comfort
The 120° adjustable finger rest – for both right- and left-handed operator comfort – allows the hand to relax during pipetting cycles.

Super Blow-out
150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action in 50μL models and below.

Wide Selection of ClipTip Pipette Tips
Choose from five sizes in 96-format: 20, 50, 200, 300, 1000μL

CE marked
The F1-ClipTip Pipette together with compatible ClipTip pipette tips is CE marked in accordance with IVD Directive in Europe.

LifetimeWarranty
With convenient online registration, and may vary by country.


Learn more about the complete ClipTip family >
Specifications

Specifications

ClipTip tips: 20, 200, 300 and 1000
F1-ClipTip GLP Kit 2: 2-20 μL, 20-200 μL, 100-1000 μL and 8-ch 30-300 μL
2 to 1000 and 30 to 300 μL
Videos
Documents