Learn More
Thermo Scientific™ EZ-Link™ Sulfo NHS-LC-LC-Biotin, No-Weigh™ Format
Brand: Thermo Scientific™ 21338
169.34 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : PN
Additional Details : Weight : 1.81440kg
Description
Thermo Scientific™ EZ-Link Sulfo-NHS-LC-LC-Biotin is a long-chain biotinylation reagent for labeling antibodies, proteins and other molecules that have primary amines.
Additional Information and Online Ordering »
Highlights:
- Forms a stable amide bond with primary amines
- Sulfonated form is water-soluble
Specifications
Sulfo-NHS-LC-LC-Biotin | |
Store desiccated at -20°C. Shipped at ambient temperature. | |
Cell-Impermeant | |
Active Ester, Succinimidyl Ester, Sulfo-NHS Ester | |
Biotin | |
Biotin & Analogs |
50 mg | |
EZ-Link™ | |
Amine | |
DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water | |
Long |
For Research Use Only. Not for use in diagnostic procedures.