Learn More
Thermo Scientific™ EZ-Link™ PFP-Biotin
Biotinylate primary amines in proteins as well as secondary amines in DNA or RNA with this very short (9.6A) pentafluorophenyl-ester biotinylation reagent.
Brand: Thermo Scientific™ 21218
Code : PN
Additional Details : CAS Number : 94481-72-8 Weight : 0.05000kg
Description
Thermo Scientific™ EZ-Link PFP-biotin is a pentafluorophenyl-ester biotinylation reagents that has similar properties to NHS-biotin but is more reactive and will target both primary and secondary amines for biotinylation.
Highlights:
- Reacts with primary and secondary amines at pH 7-9
- Pentafluorophenyl esters are more reactive than NHS-esters
- Easier to handle and store than NHS-esters
- Creates a stable amide bond
Specifications
PFP-Biotin | |
Store desiccated at 4°C. Shipped at ambient temperature. | |
Cell-Permeant | |
Active Ester, Succinimidyl Ester, TFP Ester | |
Biotin | |
Biotin & Analogs |
50 mg | |
EZ-Link™ | |
Amine | |
DMF (Dimethylformamide), DMSO (Dimethylsulfoxide) | |
Short |
For Research Use Only. Not for use in diagnostic procedures.