Learn More
Thermo Scientific™ EZ-Link™ NHS-SS-PEG4-Biotin
Biotinylate protein amines with this cleavable, long (37.9A) NHS-ester biotinylation reagent containing a reducible disulfide bond in the pegylated, water-soluble spacer arm.
Brand: Thermo Scientific™ 21442
Code : PN
Additional Details : Weight : 0.80000kg
Description
Thermo Scientific™ EZ-Link NHS-SS-PEG4-Biotin includes disulfide and polyethylene glycol (PEG) groups in its spacer arm to provide biotinylation that is cleavable and which enhances solubility.
Highlights:
- Extended spacer arm reduces steric hindrance in binding to avidin, streptavidin and Thermo Scientific NeutrAvidin Protein
- PEG spacer maintains water solubility of the modified compound
- Disulfide bond in the spacer arm enables the biotin group to be cleaved from the protein by treatment with reducing agent
- Useful in affinity purification experiments where it is necessary to recover biotin labeled protein from immobilized avidin, streptavidin or NeutrAvidin™ Supports
Specifications
NHS-SS-PEG4-Biotin | |
Store desiccated at -20°C. Shipped at ambient temperature. | |
Cell-Permeant | |
Active Ester, Succinimidyl Ester, NHS Ester | |
Biotin | |
Biotin & Analogs |
50 mg | |
EZ-Link™ | |
Amine | |
DMF (Dimethylformamide), DMSO (Dimethylsulfoxide) | |
Extra-long, Pegylated, Cleavable |
For Research Use Only. Not for use in diagnostic procedures.