missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EZ-Link™ Micro NHS-PEG4-Biotinylation Kit

Biotinylate protein amines and maximize solubility of proteins with this long (29.0A) NHS-ester biotinylation reagent containing a pegylated, water-soluble spacer arm.

Brand:  Thermo Scientific™ 21955

Code : PN

Additional Details : Weight : 3.17510kg

Product Code. 10015883

  • £283.00 / 1 set
Estimated Shipment: 26-04-2024
to see stock.
Add to basket

Description

Description

Thermo Scientific™ EZ-Link NHS-PEG&subscript_4;-Biotin is a pegylated, water-soluble reagent for simple and efficient biotin labeling of antibodies, proteins and other primary amine-containing macromolecules.

Highlights:

  • Water-soluble polyethylene glycol (PEG) spacer arm increases solubility of labeled protein
  • Prevents aggregation of antibodies better than Sulfo-NHS-LC-Biotin
  • Labels at N-terminus or lysine residues in less than 1 hour
  • Thermo Scientific No-Weigh Packaging (8 x 2mg) saves time - preserves reagent activity and prevents cross-contamination between use
Specifications

Specifications

EZ-Link Micro NHS-PEG4-Biotinylation Kit
8 Reaction Kit
100 μg
Water
Pegylated
Micro NHS-PEG4-Biotinylation Kit
Sufficient For: 8 labeling reactions, each with 0.05–0.2 mg of antibody
• EZ-Link NHS-PEG4-Biotin, 8 x 2 mg
• PBS Pack (makes 500 mL), 1 pack
• Zeba Spin Desalting Columns (7K MWCO), 2 mL, 10 columns

Store microtubes of biotin reagent at -20°C. Store PBS at room temperature. Store desalting column at 4°C. Shipped at ambient temperature.
EZ-Link™
Biotin
Antibodies, Proteins
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.