Learn More
Thermo Scientific™ EZ-Link™ Maleimide-PEG2-Biotin
Biotinylate reduced cysteines with this short, maleimide-activated, sulfhydryl-reactive biotinylation reagent containing a 2-unit PEG spacer arm to enhance water solubility.
Brand:  Thermo Scientific™ 21902BID
Code : A0
Additional Details : Weight : 2.72160kg
 View more versions of this product
Product Code. 11891255
  • £203.32 / Pack of 8
EU Stock 7
Add to basket
Description
Description
Highlights:
  • Water-soluble; contains polyethylene glycol (PEG) spacer arm
  • Reacts with sulfhydryl (-SH) groups at acidic to neutral pH
  • Transfers hydrophilic PEG spacer arm to final conjugate
  • Forms a thioether bond
Specifications
Specifications
Sulfhydryl
EZ-Link Maleimide-PEG2-Biotin, No-Weigh Format
525.62
Yes
8 x 2mg
Yes
No
Maleimide-activated biotin with diethyleneglycol spacer
No
29.1
8 uses to label a total of 203mg of IgG using typical conditions
Videos