Learn More
Thermo Scientific™ EZ-Link™ Amine-PEG4-Desthiobiotin, No-Weigh™ Format
Thermo Scientific™ EZ-Link™ Amine-PEG4-Desthiobiotin No-Weigh™ Format is a pegylated amine compound whose biotin-like group is elutable from streptavidin, making it ideal for carbodiimide (EDC) conjugation to carboxylates for labeling and purification.
Brand: Thermo Scientific™ A39263
Additional Details : Weight : 0.05200kg
Description
- Pegylated amine compound whose biotin-like group is elutable from streptavidin, making it ideal for carbodiimide (EDC) conjugation to carboxylates for labeling and purification.
- Desthiobiotin - analog of biotin allows easy elution from streptavidin, an ideal feature for affinity purification applications
- Amine-activated - primary amine can be crosslinked to proteins and material surfaces using EDC and other crosslinkers
- Pegylated - polyethylene glycol (PEG) groups in spacer arm enhances water solubility of biotinylated molecules
Specifications
Amine-PEG4-Desthiobiotin | |
Store at -20°C. | |
Wet Ice | |
Cell-Impermeant | |
Amine, Primary Amine | |
Desthiobiotin |
5 x 1 mg | |
Chemical Labeling | |
EZ-Link™ | |
Aldehyde, Carboxylic Acid, Ketone | |
Water | |
Pegylated, 28.8 Å |
For Research Use Only. Not for use in diagnostic procedures.