Learn More
Fisherbrand™ External Probe
Fisherbrand External Probe is for use with Isotemp and Isotemp Advanced hotplates and hotplate stirrers. Optimizes performance and flexibility.
£38.05 - £53.80
Specifications
Material Stainless Steel
Temperature (Metric) 0°C to 400°C
View More Specs
Products 2
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
15303548 View Documents Fisherbrand™
Isotemp (±1.8°C at 70°C) and Isotemp Advanced (±1.2°C at 70°C) hotplates and hotplate stirrers £38.05
Each View alternative products
Save Up to 
»
In Stock 7
15313548 View Documents Fisherbrand™
Isotemp (±1.0°C at 70°C) and Isotemp Advanced (±0.5°C @ 70°C) hotplates and hotplate stirrers £53.80
Each View alternative products
Save Up to 
»
In Stock 2
Description
Description
  • Optimize performance and flexibility
Specifications
Specifications
Stainless Steel
0°C to 400°C