Learn More
Thermo Scientific Expression Vectors for Pierce™ qIP Kits
Optimize cloning with this group of vectors, which includes epitope-tagged (HA and c-Myc) and TurboLuc-tagged (Tluc) multiple cloning site mammalian expression vectors.
Brand:  Thermo Scientific 82031
UNSPSC : 12352202
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15225713
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Specifications Description & Specifications
Specifications
0.5μg/μL DNA in 10mM Tris-HCl, pH 8.5
10μg