missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SuperScript™ EXPRESS One-Step Superscript™ qRT-PCR Kit, with premixed ROX
Next-generation, real-time qRT-PCR reagents designed to complement fast-cycling instruments. Get the turnaround time you want with the data quality you need.
£902.00 - £3590.00
Specifications
Content And Storage | EXPRESS One-Step SuperScript™ qRT-PCR SuperMixes are composed of 2 modules: an EXPRESS RT module (containing SuperScript™ III Reverse Transcriptase and RNAseOUT™) and an EXPRESS qPCR SuperMix module with ROX premixed or in a separate tube. Store at -20°C upon receipt. Once thawed, the EXPRESS qPCR SuperMix can be stored for up to 1 month at +4·C. Product is guaranteed stable for 6 months when properly stored. |
---|---|
Detection Method | Primer-probe |
Format | Tube |
PCR Method | 1-step RT-qPCR |
Polymerase | Taq DNA Polymerase |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
10574414
|
Invitrogen™
11791200 |
500 Reactions |
£902.00
Each |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10174783
|
Invitrogen™
1179101K |
2500 Reactions |
£3590.00
2500 reactions |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
EXPRESS One-Step SuperScript™ qRT-PCR SuperMix Kits incorporate proven Invitrogen enzymes now specifically tailored to deliver superior results on high-throughput, fast-cycling instruments for your probe-based qPCR experiments.
- Fast-activating, antibody-mediated Platinum™ Taq DNA Polymerase - complete, rapid activation provides the best sensitivity and reproducibility
- The One-Step SuperScript™ III Reverse Transcriptase module has a unique formulation to ensure better yield and less PCR inhibition, specifically for one-step qRT-PCR
- Flexible formats with ROX premixed or separate - designed to work with default instrument protocols on a wide range of high-throughput fast instruments (e.g., AB 7500 Fast, AB 7900 HT Fast, AB StepOne, Roche LightCycler™ 480, Corbett Rotor-Gene™)
- The only commercially available one-step qRT-PCR kits containing heat-labile UDG - significantly reduced carryover contamination risk with excellent activity from room temperature to approximately 40°C. Due to the thermostability of SuperScript™ III, the cDNA synthesis step is run at 50°C, at which temperature the heat-labile UDG is totally and irreversibly inactivated
Easy to test and switch:
- EXPRESS kits work with manufacturer's default protocols on fast instruments and come in a convenient and stable SuperMix format for ultimate convenience
- The kits are offered with either premixed ROX or a separate tube of ROX to fit your instrument needs
Order Info
Guaranteed to be stable for 6 months when properly stored.
Specifications
EXPRESS One-Step SuperScript™ qRT-PCR SuperMixes are composed of 2 modules: an EXPRESS RT module (containing SuperScript™ III Reverse Transcriptase and RNAseOUT™) and an EXPRESS qPCR SuperMix module with ROX premixed or in a separate tube. Store at -20°C upon receipt. Once thawed, the EXPRESS qPCR SuperMix can be stored for up to 1 month at +4·C. Product is guaranteed stable for 6 months when properly stored. |
|
Tube | |
Taq DNA Polymerase | |
7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System, Eppendorf™ MasterCycler™ RealPlex, StepOne™, Fast Mode, StepOne™, Standard Mode | |
qRT-PCR Kit | |
RNA |
Primer-probe | |
1-step RT-qPCR | |
Fast or Standard | |
SuperScript™ | |
Gene Expression |
For Research Use Only. Not for use in diagnostic procedures.