Learn More
Applied Biosystems™ ExoSAP-IT™ PCR Product Cleanup Reagent
ExoSAP-IT™ PCR Product Clean-up Reagent is used for enzymatic clean-up of amplified PCR product. It hydrolyses excess primers and nucleotides in a single step.
Brand: Applied Biosystems™ 78200.200.UL
Code : NEW
Additional Details : Weight : 1.00000kg
Description
- Remove excess primers and unincorporated nucleotides enzymatically
- Prepare DNA sequencing samples with one pipetting step
- Conserve precious PCR products with up to 100% recovery
- Minimize column clean-up and sample loss
Specifications
PCR Purification | |
ExoSAP-IT™ PCR Product Clean-up Reagent, 100 rxns, Recombinant, Dry Ice, DNA Sequencing or Single Nucleotide Polymorphism (SNP) Analysis, Use with PCR Purification, Store at –5 to °30°C | |
Store at -5 to -30°C. | |
Genetic Diseases | |
PCR Purification |
100 rxns | |
200μL | |
Recombinant | |
Dry Ice | |
ExoSAP-IT™ |