Learn More
Applied Biosystems™ ExoSAP-IT™ Express PCR Product Cleanup Reagent
Removes excess primers and dNTPs. Does not interfere with downstream applications.
Brand: Applied Biosystems™ 75001.4X.1.ML
Code : NEW
Additional Details : Weight : 0.47500kg
Description
- Mixture of two hydrolytic enzymes, a modified Exonuclease I and Shrimp Alkaline Phosphatase (SAP), in a specially formulated buffer
- One-tube, one-step PCR cleanup
- Five-minute protocol
- Single tube, 8-tube strip, and 96-well plate formats offer flexibility from manual pipetting to automated workflows
Specifications
-20°C | |
2000 |