Learn More
Exo1, Tocris Bioscience™
Inhibits Golgi-ER traffic; blocks exocytosis
Brand:  Tocris (Bio-Techne) 185050
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13440124
  • £358.68 / 50mg
Estimated Shipment date
from Supplier 28-10-2019
Description
Description
Reversible inhibitor of vesicular trafficking between endoplasmic reticulum and Golgi apparatus. Modifies Golgi ADP-ribosylation factor (ARF)1 GTPase activity and inhibits exocytosis (IC50 ∽ 20 mM). Has similar, but not identical, action to brefeldin A (Cat. No. 1231).
Specifications
Chemical Identifiers
461681-88-9
273.263
exo1, methyl 2-4-fluorobenzoyl amino benzoate, exo-1, 2-4-fluorobenzoylamino benzoic acid methyl ester, 2-4-fluorobenzoylamino methylbenzoate, methyl 2-4-fluorobenzamido benzoate, exo 1, exo hplc , solid, methyl 2-4-fluorophenyl carbonylamino benzoate, 2-4-fluoro-benzoylamino-benzoic acid methyl ester
methyl 2-[(4-fluorobenzoyl)amino]benzoate
C15H12FNO3
KIAPWMKFHIKQOZ-UHFFFAOYSA-N
310557
COC(=O)C1=CC=CC=C1NC(=O)C2=CC=C(C=C2)F
Specifications
Exo1
461681-88-9
exo1, methyl 2-4-fluorobenzoyl amino benzoate, exo-1, 2-4-fluorobenzoylamino benzoic acid methyl ester, 2-4-fluorobenzoylamino methylbenzoate, methyl 2-4-fluorobenzamido benzoate, exo 1, exo hplc , solid, methyl 2-4-fluorophenyl carbonylamino benzoate, 2-4-fluoro-benzoylamino-benzoic acid methyl ester
COC(=O)C1=CC=CC=C1NC(=O)C2=CC=C(C=C2)F
273.263
273.26
50mg
C15H12FNO3
KIAPWMKFHIKQOZ-UHFFFAOYSA-N
methyl 2-[(4-fluorobenzoyl)amino]benzoate
310557
>99%