missing translation for 'onlineSavingsMsg'
Learn More

exo-IWR 1, Tocris Bioscience™

Negative control for endo-IWR 1 (Cat. No. 3532)

Brand:  Tocris (Bio-Techne) 394710

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13463864

  • £137.70 / 10mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Negative control for endo-IWR 1 (Cat.No. 3532). 25-fold less active than endo-IWR 1; exhibits decreased activity against the Wnt/β-catenin pathway.
Specifications

Chemical Identifiers

1127442-87-8
409.445
iwr-1-exo, exo-iwr 1, 4-3ar,4r,7s,7as-1,3-dioxo-1,3,3a,4,7,7a-hexahydro-2h-4,7-methanoisoindol-2-yl-n-quinolin-8-yl benzamide, exo-iwr-1, 4-3ar,4r,7s,7as-rel-1,3,3a,4,7,7a-hexahydro-1,3-dioxo-4,7-methano-2h-isoindol-2-yl-n-8-quinolinylbenzamide, rel-4-3ar,4r,7s,7as-1,3,3a,4,7,7a-hexahydro-1,3-dioxo-4,7-methano-2h-isoindol-2-yl-n-8-quinolinylbenzamide
CHEBI:62883
C25H19N3O3
ZGSXEXBYLJIOGF-BTYSMDAFSA-N
1163034
C1C2C=CC1C3C2C(=O)N(C3=O)C4=CC=C(C=C4)C(=O)NC5=CC=CC6=C5N=CC=C6

Specifications

exo-IWR 1
1127442-87-8
iwr-1-exo, exo-iwr 1, 4-3ar,4r,7s,7as-1,3-dioxo-1,3,3a,4,7,7a-hexahydro-2h-4,7-methanoisoindol-2-yl-n-quinolin-8-yl benzamide, exo-iwr-1, 4-3ar,4r,7s,7as-rel-1,3,3a,4,7,7a-hexahydro-1,3-dioxo-4,7-methano-2h-isoindol-2-yl-n-8-quinolinylbenzamide, rel-4-3ar,4r,7s,7as-1,3,3a,4,7,7a-hexahydro-1,3-dioxo-4,7-methano-2h-isoindol-2-yl-n-8-quinolinylbenzamide
C1C2C=CC1C3C2C(=O)N(C3=O)C4=CC=C(C=C4)C(=O)NC5=CC=CC6=C5N=CC=C6
1163034
409.44
10mg
C25H19N3O3
ZGSXEXBYLJIOGF-BTYSMDAFSA-N
409.445
CHEBI:62883
>99%
Videos