missing translation for 'onlineSavingsMsg'
Learn More

exo-2-Aminonorbornane, 99%, ACROS Organics™

Brand:  Acros Organics 176950050

Code : 29

Additional Details : CAS Number : 7242-92-4 Weight : 0.05500kg Transport : UN number : 2733 Chem class : 3 Pack group : III


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10764862

  • £114.40 / 5g


Estimated Shipment
18-06-2018
Add to basket


Specifications

Chemical Identifiers

exo-2-Aminonorbornane
99%
111.19
99%
C7H13N
exo-2-Norbornanamine

Specifications

exo-2-Aminonorbornane
Authentic
1.4807
49.0°C (13.0 mbar)
Liquid
99%
35°C
111.19
7242-92-4
Glass bottle
0.9300g/mL
Clear colorless to pale yellow
99%
5g
C7H13N
exo-2-Norbornanamine
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • exo-2-Aminonorbornane

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS