Learn More
EVOS™ Imaging System Accessory, Microscope Objectives
Components manufactured to ensure publication-quality images
Brand:  EVOS™ AMEP4633
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11987275
  • POA / Each
This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on 01509 555500 for assistance.
Specifications
Specifications
EVOS™ digital microscope
EVOS™ LWD achromatic phase contrast objective
0.25
10X
Videos