Learn More
EVOS™ Imaging System Accessory, Microscope Objectives
Components manufactured to ensure publication-quality images
Brand:  EVOS™ AMEP4631
Code : NEW
Additional Details : Weight : 0.01000kg
Product Code. 11967275
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
RMS-threaded objectives with 45mm parfocal distance