Learn More
EVOS™ Imaging System Accessory, Microscope Objectives
Components manufactured to ensure publication-quality images
Brand:  EVOS™ AMEP4634
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11997275
  • £698.68 / Each
Estimated Shipment
21-05-2018
Add to basket
Description
Description
RMS-threaded objectives with 45mm parfocal distance
Specifications
Specifications
EVOS™ digital microscope
EVOS™ LWD achromatic phase contrast objective
0.40
20X
Videos