Learn More
EVOS™ Light Cubes
Light engine outputs remarkable intensity over a short light path that delivers superior fluorophore excitation
Brand:  EVOS™ AMEP4656
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11977285
  • £2689.48 / Each
Estimated Shipment
18-05-2018
Add to basket
Description
Description
  • Ideal for use with EVOS FL Color Imaging System, EVOS FL Auto Imaging System or EVOS FL Imaging Systems
  • Each cube contains an LED, condensing optics, and hard-coated filters
  • Continuous illumination enables consistent results 50,000+ hour bulb lifetime that affords lower operating costs
  • Adjustable light intensity that helps reduce photobleaching User changeable and automatically recognized by the system
Specifications
Specifications
EVOS™ digital microscope
CY5 Fluorescent light cube
Videos