Learn More
EVOS™ Light Cubes
Light engine outputs remarkable intensity over a short light path that delivers superior fluorophore excitation
Brand:  EVOS™ AMEP4655
Code : A0
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11967285
  • £2699.84 / Each
Estimated Shipment
07-06-2018
Add to basket
Description
Description
  • Ideal for use with EVOS FL Color Imaging System, EVOS FL Auto Imaging System or EVOS FL Imaging Systems
  • Each cube contains an LED, condensing optics, and hard-coated filters
  • Continuous illumination enables consistent results 50,000+ hour bulb lifetime that affords lower operating costs
  • Adjustable light intensity that helps reduce photobleaching User changeable and automatically recognized by the system
Specifications
Specifications
EVOS™ digital microscope
Texas red Fluorescent light cube
Videos