missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Evolution™ 260 Bio UV-Visible Spectrophotometer

The Thermo Scientific™ Evolution 260 spectrophotometer offers reliable, high-quality performance and enhanced customer usability.

Brand:  Thermo Scientific™ 840-210900

Code : 04

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10736369

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

Complete Life Science Applications

Next generation INSIGHT software provides the streamlined convenience of easy-to-understand nucleic acid and protein analysis methods and includes dedicated modules for:

  • Nucleic Acid concentration by direct UV measurement at 260nm including DNA, ssDNA, RNA, and siRNA
  • Nucleic acid purity measurements from 260/280nm and 260/230 ratios
  • Protein analysis by direct UV measurement at 205 or 280nm
  • Most common colorimetric analysis methods including: Pierce 660nm Protein assay, Bradford, BCA and Lowry
  • Measuring the fluorescence labeling efficiency for proteins and nucleic acids
  • Measuring the microarray labeling efficiency with one or two dyes

Customizable, Straightforward INSIGHT Software

  • Personalize your home screen for easy navigation and convenience
  • Workbook tools save time and ensure accurate results
  • CUE scripting allows creation of customized interface and simplified workflow

Best-in-Class Performance

  • Double-beam geometry ideal for kinetics or any sample that may change over time
  • Application Focused Beam Geometry (AFBG) technology provides superior results for microcells and turbid samples
  • Xenon flash lamp requires no warm-up time allowing instant measurements
  • Electronic triggering provides the highest level of accuracy for rapid-mixing kinetics measurements
  • Precision monochromator drive delivers fast scanning data collection and wavelength accuracy Powerful Convenience at your Fingertips
  • Color, touch screen provides instrument control using built-in computer
  • Integrated keypad allows communication with INSIGHT software
  • Choice of Local or PC configurations provides flexibility and convenience

Accessories for all your Sampling Needs

  • Wide accessory selection with various temperature control options, sample changers, sippers and solid sampling devices complete your analysis
  • Automated cell changers with Peltier temperature control increase sample throughput
  • Choice of Peltier Single Cell Holder or a Smart 8-Cell Peltier system for temperature control and sample monitoring from 0° to 100°C
  • Easy software integration gives you complete control of your measurements

Reliable Assurance of System Performance

  • The Thermo Scientific UV Validator package provides support for system qualification and validation activities
  • Complete documentation facilitates compliance of your system with the requirements of FDA, GxP, ISO 9001:2000 and ISPE 2001 guidelines and regulations
  • CVC and Mercury Lamp accessory automated testing ensures accuracy of your data
  • Thermo Scientific INSIGHTsecurity software provides complete tools to meet 21 CFR Part 11 requirements
Specifications

Specifications

1A: ±0.006A
2A: ±0.010A
Measured at 440nm using calibrated neutral density filters traceable to NIST
Double-beam with Application Focused Beam Geometry
10, 5, 2, 1.0, 0.5, 0.2, 0.1nm
Dual Silicon Photodiodes
Touchscreen LCD panel; 800 x 480; 17.8 cm (7 in. diagonal)
100-240 V 50-60 Hz selected automatically150W maximum
Sealed membrane
0A: <0.00015A
1A: <0.00025A
2A: <0.00080A
260nm, 1.0nm SBW, RMS
Double Beam with sample and reference cuvette positions; Application Focused Beam Geometry; Czerny-Turner Monochromator
1A: ±0.006A
2A: ±0.010A
Measured at 440nm using calibrated neutral density filters traceable to NIST
>3.5A
-
Absorbance, % Transmittance, % Reflectance, Kubelka-Munk, log (1/R), log (Abs), Abs*Factor, Intensity
Variable: 1.0nm; 2.0nm; AFBG microcell optimized; AFBG fiber optic optimized; AFBG materials optimized
±0.5nm (541.9, 546.1nm mercury lines)
±0.8nm (full range 190-1100nm)
<1 to 6,000 nm/min; variable
14.4 kg
±0.0010A, 200-800nm, 1.0nm SBW, smoothing
ISO 9001:2008
Evolution 260 Bio Local Control Spectrophotometer
62.2 x 48.6 x 27.9 cm
<0.0005 A/hr, 500 nm, 1.0nm SBW, 1 hr warm-up
Evolution 260 LC Spectrophotometer
Xenon Flash Lamp, 3-year warranty (7 years typical lifetime)
Microsoft Windows™ XP embedded
INSIGHT 2 Software is also compatible with Windows 7 and Windows 8.1 Professional Editions and can be installed on a computer attached to the instrument
(Guaranteed Performance Specifications)
Resolution (toluene in Hexane): ≥1.8A
Photometric Accuracy (60mg/L K2Cr2O7): ±0.010A
Stray Light: ≤1%T at 198nm: KCI; ≤0.05%AT at 220nm: Nal, KI
Wavelength Accuracy: ±0.5nm 541.9, 546.1nm Hg emission lines, ±0.8nm full range
Wavelength Repeatability: ≤0.05nm, repetitive scanning of 546.1nm Hg emission line
-0.3 - 4.0A
±0.0002A
-
<1-6000nm/min, variable
Evolution 260 Bio Local Control Spectrophotometer
190 - 1,100nm
-
Videos
Documentation