missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EVI2B Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-99056UV
This item is not returnable.
View return policy
Description
EVI2B Polyclonal specifically detects EVI2B in Mouse samples. It is validated for Western Blot.Specifications
EVI2B | |
Polyclonal | |
Western Blot | |
CD361 antigen, D17S376, ecotropic viral integration site 2B, Ecotropic viral integration site 2B protein homolog, EVDBprotein EVI2B, EVI-2B | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of mouse EVI2B. | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Western Blot | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
2124 | |
Store at 4°C in the dark. | |
IgG |