missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Evaporating Capsules

Use these dishes for a variety of applications. Haldenwanger™ Evaporating Capsules feature a smooth internal finish, constructed from optimized laboratory porcelain.

Brand:  Haldenwanger™ 131/4

Code : 02

Additional Details : Weight : 0.06500kg

 View more versions of this product

Product Code. 12085053

  • £19.19 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

Flat
84mm
36mm
Evaporating Capsule
100mL
84mm
Porcelain
Videos