missing translation for 'onlineSavingsMsg'
Learn More

EVA1, PE, clone: G9P3-1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-9742-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15527226

  • £68.96 / 25µg


Estimated Shipment date
from Supplier 04-06-2018
Add to basket

Description

Description

Description: The monoclonal antibody G9P3-1 reacts with human and mouse epithelial V-like antigen 1 (EVA1), also known as myelin protein zero-like 2 (MPZL2). EVA1 is a low affinity adhesion type I transmembrane glycoprotein that bears an immunolobulin V-type domain. This glycoprotein is expressed on some epithelial tissues and also on immature thymocytes in the DN3 stage. The DN3 thymocytes are characterized by CD4 and CD8 double negative cells that are CD25 positive and CD44 negative. EVA1 has a role in immature thymocyte development, and thymic regeneration.Applications Reported: This G9P3-1 antibody has been reported for use in flow cytometric analysis.Applications Tested: This G9P3-1 antibody has been tested by flow cytometric analysis of mouse thymocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

EVA1
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q9H8M9, Q91WM6
Transmembrane protein 166
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
G9P3-1
PE
EVA1A
Conjugated
EVA1A
IgG1, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
84141
Videos

For Research Use Only.