Learn More
Thermo Scientific™ Eutech™ Ion Selective Combination Electrode, Epoxy-Body
Use Eutech™ Ion Selective Combination Electrode for measurement of specific ions in aqueous solutions.
Brand: Thermo Scientific™ CAL0301B
Code : 04
Additional Details : Weight : 0.10000kg
Description
Eutech™ offers a wide selection of Ion Selective Electrodes (ISE) and meters for the measurement of ammonia, bromide, cadmium, calcium, carbon dioxide, carbonate, chloride, copper, cyanide, fluoride, iodide, lead, nitrate, potassium, silver, sulfide and sodium.ISE responds to ionic activity or concentration in a liquid because of the characteristics of the electrode’s sensing membrane. The ISE develops an electrical potential which is proportional to the activity or concentration of the ion for which the membrane is selective.
Features:
- Epoxy-body
- For Use with: IS meters
Specifications
Epoxy-body | |
Calcium | |
Eutech™ Ion Meters | |
Calcium |