missing translation for 'onlineSavingsMsg'
Learn More

IKA™ EUROSTAR 200 Control P4 Overhead Stirrer

Extremely powerful laboratory stirrer for highly viscous applications for quantities up to 100 L (H2O).

Brand:  IKA™ 0004090000

Code : A0

Additional Details : Weight : 4.00000kg

 View more versions of this product

Product Code. 13440577

  • £4522.91 / Each


Estimated Shipment
17-03-2020
Add to basket

Description

Description

  • Designed with a removable wireless controller and a digital TFT display
  • Automatically adjusts the speed through microprocessor controlled technology within the speed range
  • The stirrer comes equipped with a RS 232 and a USB interface to control and document all parameters
  • An integrated torque trend display is provided for the measurement of viscosity changes
  • Safety circuits installed ensures automatic cut-off in an anti-stall or overload conditions
  • Continuous comparison of shaft speed to desired speed is maintained and variations are adjusted automatically
  • Guarantees a constant speed even with changes in viscosities of the sample
  • Multilingual TFT display
  • Programmable functions
  • Integrated temperature measurement
  • Interval operation
  • Timer function
  • Adjustable safety circuit
  • Locked function
  • Infinitely adjustable speed
  • Overload protection
  • Short-term overload operation
  • Slim casing
  • Quiet operation
  • Error code display
  • H 67.60 temperature sensor and WH 11 WiCo holder included in delivery
Specifications

Specifications

TFT
4 to 530RPM
379mm
EUROSTAR 200 Control P4
230V
91mm
660Ncm
230V, 50/60Hz, 134w
50/60Hz
5 to 40°C
Alu-Cast Coating/Thermoplastic Polymer
4 to 530 rpm
134w
150000mPas
100 L
EU Plug
Videos
Documents