missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Europium ingot, 99.9% (REO), Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under mineral oil
Specifications
Color | Gray |
UN Number | UN3208 |
Quantity | 1 g |
Percent Purity | 99.9% |
Odor | Odorless |
Form | Ingot |
Assay | (REO) |
Chemical Name or Material | Europium ingot |
Safety and Handling
Product Identifier
- Europium ingot
- Danger
- Pyrophoric solid Category 1
- Substance which\, in contact with water\, emits flammable gases Category 2
- H250-Catches fire spontaneously if exposed to air.
- H261-In contact with water releases flammable gases.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P222-Do not allow contact with air.
- P223-Do not allow contact with water.
- P231+P232-Handle under inert gas. Protect from moisture.
- P422-Store contents under .
- P501-Dispose of contents/container to .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'