missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) trifluoroacetate hydrate, 99.9% (metals basis), Thermo Scientific™

Catalog Number p-7059454
Click to view available options
Quantity:
50 g
This item is not returnable. View return policy
Eu(OOCCF3)3·xH2O
MFCD05663782
This item is not returnable. View return policy
Eu(OOCCF3)3·xH2O
MFCD05663782

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Physical Form Crystalline
Quantity 50 g
UN Number UN3288
Solubility Information Soluble in most polar and non polar solids.
Formula Weight 490.97 (Anhydrous)
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Europium (III) trifluoroacetate hydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'