missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Europium(III) oxide, 99.9% (REO), Thermo Scientific Chemicals
103.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
2 g
10 g
Description
Europium(III) oxide is used for vacuum deposition, fluorescent lamps, and as an activator for yttrium-based phosphors, manufacture of fluorescent glass, anti-counterfeiting phosphors in Euro banknotes. And it is widely used as a red or blue phosphor in television sets.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 2291°C |
Physical Form | Sintered Lump |
Quantity | 2 g |
Merck Index | 14,3904 |
Solubility Information | Soluble in acids. Insoluble in water. |
IUPAC Name | dieuropium(3+) trioxidandiide |
Formula Weight | 351.92 |
Percent Purity | 99.9% |
Sensitivity | Hygroscopic |
Assay | (REO) |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'