missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Europium(III) chloride hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Europium(III) chloride hydrate is used as a catalyst in organic synthesis. It is involved in the preparation of amino acid based europium(III) complex in order to prepare photoactive thin films, which finds application in organic light emitting diodes. It is also used as a precursor for the preparation of other europium compounds and its salts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Yellow |
Physical Form | Crystalline |
Quantity | 1 g |
Assay Percent Range | 99.99% (REO) |
Solubility Information | Soluble in water. |
Formula Weight | 258.32 (Anhydrous) |
Odor | Odorless |
Sensitivity | Hygroscopic |
Appearance | Yellow crystalline |
Chemical Name or Material | Europium(III) chloride hydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'