Learn More
Thermo Scientific Alfa Aesar Europium foil, 0.1mm (0.004 in.) thick, 99.9% (REO)
Brand: Thermo Scientific Alfa Aesar 013965.FI
Europium foil, 0.1mm (0.004 in.) thick, 99.9% (REO)
Code : NEW
Additional Details : CAS Number : 7440-53-1 Weight : 0.01000kg Transport : UN number : 3208 Chem class : 4.3 Pack group : II