missing translation for 'onlineSavingsMsg'
Learn More

Europium, AAS standard solution, Specpure™ Eu 1000μg/mL, Thermo Scientific Chemicals

Catalog Number p-7080407
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy
MFCD00010992
This item is not returnable. View return policy
MFCD00010992

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Concentration Eu2O3 in 5% HNO3
Quantity 100 mL
UN Number UN3264
Grade Specpure™
Physical Form Liquid
Chemical Name or Material Europium, AAS standard solution, Eu 1000μg/mL

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'