missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETS1 Mouse, Clone: ETS1/1282, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 2113-MSM1-P1
Additional Details : Weight : 0.01000kg
Specifications
ETS1 | |
Nuclear | |
ETS1/1282 | |
Unconjugated | |
NP_001155894 | |
Mouse | |
54kDa | |
Protein A/G Purified | |
RUO | |
2113 | |
IgG |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
ETS1 | |
Recombinant human ETS1 fragment (aa137-230) | |
HepG2 and HeLa cells. Tonsil, Skin, Pancreas, Kidney and Lymphoma. | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |