missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETS1 Mouse, Clone: ETS1/1282, enQuireBio™
Mouse Monoclonal Antibody
£181.83 - £395.00
Specifications
Antigen | ETS1 |
---|---|
Clone | ETS1/1282 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15949967
|
EnquireBio
2113-MSM1-P1 |
100 μg |
£395.00
100µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15939967
|
EnquireBio
2113-MSM1-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
ETS1 | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
2113 | |
Recombinant human ETS1 fragment (aa137-230) | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
54kDa |
ETS1/1282 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
NP_001155894 | |
ETS1 | |
IgG | |
Protein A/G Purified | |
Nuclear | |
HepG2 and HeLa cells. Tonsil, Skin, Pancreas, Kidney and Lymphoma. |