missing translation for 'onlineSavingsMsg'
Learn More

Etifoxine hydrochloride, Tocris Bioscience™

Ligand of the translocator protein (TSPO); also GABAA receptor potentiator

Brand:  Tocris (Bio-Techne) 311350

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13432464

  • £520.38 / 50mg

Estimated Shipment date
from Supplier 01-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potentiator of GABAA receptor function in cultured neurons. Preferentially acts on β2 or β3 subunit-containing GABAA receptors. Exhibits anxiolytic activity in rodents and humans with no sedative, myorelaxant or mnesic side effects. Acts as a ligand of the translocator protein (TSPO); promotes axonal regeneration.
Specifications

Chemical Identifiers

56776-32-0
337.244
etifoxine hydrochloride, stresam, 6-chloro-n-ethyl-4-methyl-4-phenyl-4h-3,1-benzoxazin-2-amine monohydrochloride, etifoxine hcl, 6-chloro-2-n-ethyl-4-methyl-4-phenyl-4h-3,1-benzoxazin-2-amine hydrochloride, c17h17cln2o.clh, stresam tn, etifoxin hydrochloride, etifoxine hydrochloride hplc
6-chloro-N-ethyl-4-methyl-4-phenyl-3,1-benzoxazin-2-amine;hydrochloride
C17H18Cl2N2O
SCBJXEBIMVRTJE-UHFFFAOYSA-N
171544
CCNC1=NC2=C(C=C(C=C2)Cl)C(O1)(C)C3=CC=CC=C3.Cl

Specifications

50mg
56776-32-0
etifoxine hydrochloride, stresam, 6-chloro-n-ethyl-4-methyl-4-phenyl-4h-3,1-benzoxazin-2-amine monohydrochloride, etifoxine hcl, 6-chloro-2-n-ethyl-4-methyl-4-phenyl-4h-3,1-benzoxazin-2-amine hydrochloride, c17h17cln2o.clh, stresam tn, etifoxin hydrochloride, etifoxine hydrochloride hplc
CCNC1=NC2=C(C=C(C=C2)Cl)C(O1)(C)C3=CC=CC=C3.Cl
337.244
337.24
Etifoxine hydrochloride
C17H18Cl2N2O
SCBJXEBIMVRTJE-UHFFFAOYSA-N
6-chloro-N-ethyl-4-methyl-4-phenyl-3,1-benzoxazin-2-amine;hydrochloride
171544
>99%
Videos
Documents