missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethyltriphenylphosphonium acetate, 70% in methanol

Brand:  Alfa Aesar™ 017426.22

Code : NEW

Additional Details : CAS Number : 35835-94-0 Weight : 0.10000kg Transport : UN number : 1992 Chem class : 3 Pack group : II

Quantity 100g

 View more versions of this product

Product Code. 11363347

  • £30.21 / 100g


Estimated Shipment date
from Supplier 31-05-2018
Add to basket

Specifications

Chemical Identifiers

Ethyltriphenylphosphonium acetate
CH3CH2P(C6H5)3O2CCH3
MFCD00061389
35835-94-0
350.4
UN1993

Specifications

Ethyltriphenylphosphonium acetate
Liquid
350.4
UN1993
Yellow to dark brown
100g
35835-94-0
CH3CH2P(C6H5)3O2CCH3
MFCD00061389
1.05g/mL
70% in methanol
−17° (0°F)
Videos
SDS