missing translation for 'onlineSavingsMsg'
Learn More

Ethylphosphonic acid, 98+%, Thermo Scientific Chemicals

Catalog Number p-7026526
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
6779-09-5
C2H5O3P
108.03
MFCD00002141
GATNOFPXSDHULC-UHFFFAOYSA-L
This item is not returnable. View return policy
6779-09-5
C2H5O3P
108.03
MFCD00002141
GATNOFPXSDHULC-UHFFFAOYSA-L

Ethylphosphonic acid is used as an internal standard in the determination of fosfomycin in human plasma. It is also used in the synthesis of nucleotide analogs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylphosphonic acid is used as an internal standard in the determination of fosfomycin in human plasma. It is also used in the synthesis of nucleotide analogs.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and bases.
TRUSTED_SUSTAINABILITY
Melting Point 58°C to 62°C
Quantity 1 g
UN Number UN3261
Beilstein 1743032
Synonym styrylboronic acid, e-styrylboronic acid, e-phenylethenylboronic acid, trans-2-phenylvinylboronic acid, e-2-phenylethenyl boronic acid, 2-phenylvinylboronic acid, trans-phenylethenylboronic acid, trans-phenylvinylboronic acid, e-styreneboronic acid
SMILES CCP([O-])([O-])=O
IUPAC Name [(E)-2-phenylethenyl]boronic acid
PubChem CID 5702628
Formula Weight 110.06
Percent Purity ≥98%
Chemical Name or Material Ethylphosphonic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'