missing translation for 'onlineSavingsMsg'
Learn More

Ethylphosphonic acid, 98+%, Thermo Scientific Chemicals

Catalog Number 11415137
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
6779-09-5
C2H5O3P
108.03
MFCD00002141
GATNOFPXSDHULC-UHFFFAOYSA-L
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
6779-09-5
C2H5O3P
108.03
MFCD00002141
GATNOFPXSDHULC-UHFFFAOYSA-L

Ethylphosphonic acid is used as an internal standard in the determination of fosfomycin in human plasma. It is also used in the synthesis of nucleotide analogs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylphosphonic acid is used as an internal standard in the determination of fosfomycin in human plasma. It is also used in the synthesis of nucleotide analogs.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and bases.
TRUSTED_SUSTAINABILITY
Melting Point 58°C to 62°C
Quantity 5 g
UN Number UN3261
Beilstein 1743032
Sensitivity Hygroscopic
Synonym phosphonic acid, ethyl, ethylphosphonate, ethanephosphonic acid, phosphate analogue, 6, 4-ethylphosphonic acid, 2-propan-4-ylbenzoic acid, ethylphosphonic acid
Solubility Information Soluble in water.
SMILES CCP(=O)(O)O
IUPAC Name ethylphosphonic acid
PubChem CID 204482
Formula Weight 110.06
Percent Purity ≥98%
Chemical Name or Material Ethylphosphonic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'