missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylphosphonic acid, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Ethylphosphonic acid is used as an internal standard in the determination of fosfomycin in human plasma. It is also used in the synthesis of nucleotide analogs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethylphosphonic acid is used as an internal standard in the determination of fosfomycin in human plasma. It is also used in the synthesis of nucleotide analogs.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents and bases.
Ethylphosphonic acid is used as an internal standard in the determination of fosfomycin in human plasma. It is also used in the synthesis of nucleotide analogs.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents and bases.
Specifications
Melting Point | 58°C to 62°C |
Quantity | 1 g |
UN Number | UN3261 |
Beilstein | 1743032 |
Synonym | styrylboronic acid, e-styrylboronic acid, e-phenylethenylboronic acid, trans-2-phenylvinylboronic acid, e-2-phenylethenyl boronic acid, 2-phenylvinylboronic acid, trans-phenylethenylboronic acid, trans-phenylvinylboronic acid, e-styreneboronic acid |
SMILES | CCP([O-])([O-])=O |
IUPAC Name | [(E)-2-phenylethenyl]boronic acid |
PubChem CID | 5702628 |
Formula Weight | 110.06 |
Percent Purity | ≥98% |
Mehr anzeigen |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'